4-Tetrahydrothiopyranone-1,1-dioxide
Supplier: Matrix Scientific
Synonyms:
Tetrahydrothiopyran-4-one 1,1-Dioxide, 4-Tetrahydrothiopyranone 1,1-dioxide, 4-Oxothiane 1,1-Dioxide
062430-500MG
062430-1G
101923-946EA
52.31
USD
101923-946
102833-314
4-Tetrahydrothiopyranone-1,1-dioxide
4-Tetrahydrothiopyranone-1,1-dioxide
Hazard: Irritant
Formula:
C₅H₈O₃S MW: 148.1802 g/mol |
MDL Number:
MFCD00460698 CAS Number: 17396-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...