5-Formyl-2-methylbenzonitrile ≥95%
Supplier: Matrix Scientific
061397-1G
061397-5G
101921-888EA
256.92
USD
101921-888
101921-890
5-Formyl-2-methylbenzonitrile ≥95%
5-Formyl-2-methylbenzonitrile
Hazard: Irritant
Formula:
C₉H₇NO |
MDL Number:
MFCD12197728 CAS Number: 27613-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...