Trimethoprim

Supplier: Matrix Scientific

Synonyms: 5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine

058373-5G 058373-25G 058373-100G
101914-678EA 15.38 USD
101914-678 101914-680 101914-682
Trimethoprim
Trimethoprim

Hazard: Irritant

Formula: C₁₄H₁₈N₄O₃
MW: 290.32 g/mol
MDL Number: MFCD00036761
CAS Number: 738-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR