(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

058271-500MG 058271-1G
101914-480EA 735.38 USD
101914-480 101914-482
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanamine hydrochloride ≥95%
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClN₃
MDL Number: MFCD17167332
CAS Number: 1246738-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR