2-[3-(Trifluoromethyl)phenyl]acetohydrazide ≥95%

Supplier: Matrix Scientific

058270-500MG 058270-1G 058270-5G
101914-474EA 367.69 USD
101914-474 101914-476 101914-478
2-[3-(Trifluoromethyl)phenyl]acetohydrazide ≥95%
2-[3-(Trifluoromethyl)phenyl]acetohydrazide

Hazard: Irritant

Formula: C₉H₉F₃N₂O
MDL Number: MFCD11643284
CAS Number: 116622-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR