2-(1H-1,2,4-Triazol-1-yl)ethan-1-ol ≥95%

Supplier: Matrix Scientific

058269-500MG 058269-1G
101914-470EA 500 USD
101914-470 101914-472
2-(1H-1,2,4-Triazol-1-yl)ethan-1-ol ≥95%
2-(1H-1,2,4-Triazol-1-yl)ethan-1-ol

Hazard: Irritant

Formula: C₄H₇N₃O
MDL Number: MFCD00607979
CAS Number: 3273-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR