tert-Butyl-N-(1,3-thiazol-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl thiazol-2-ylcarbamate

058268-500MG 058268-1G
101914-466EA 278.46 USD
101914-466 101914-468
tert-Butyl-N-(1,3-thiazol-2-yl)carbamate ≥95%
tert-Butyl-N-(1,3-thiazol-2-yl)carbamate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD09038327
CAS Number: 170961-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR