tert-Butyl-4-(5-fluoro-6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

058267-500MG 058267-1G
101914-462EA 747.69 USD
101914-462 101914-464
tert-Butyl-4-(5-fluoro-6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(5-fluoro-6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄FN₃O₃
MW: 349.41 g/mol
MDL Number: MFCD17167316
CAS Number: 932374-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR