tert-Butyl-N-[1-(3-aminophenyl)ethyl]carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl [1-(3-aminophenyl)ethyl]carbamate

058264-500MG 058264-1G
101914-452EA 907.69 USD
101914-452 101914-454
tert-Butyl-N-[1-(3-aminophenyl)ethyl]carbamate ≥95%
tert-Butyl-N-[1-(3-aminophenyl)ethyl]carbamate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD06660373
CAS Number: 180079-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR