tert-Butyl-4-amino-4-carbamoylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1-Boc-piperidine-4-carboxamide, tert-Butyl 4-amino-4-carbamoylpiperidine-1-carboxylate

058262-500MG 058262-1G
101914-444EA 1296.92 USD
101914-444 101914-446
tert-Butyl-4-amino-4-carbamoylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-amino-4-carbamoylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₁N₃O₃
MW: 243.31 g/mol
MDL Number: MFCD04114477
CAS Number: 288154-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR