1-[(tert-Butoxy)carbonyl]-2-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

058261-500MG 058261-1G
101914-440EA 1084.62 USD
101914-440 101914-442
1-[(tert-Butoxy)carbonyl]-2-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylic acid ≥95%
1-[(tert-Butoxy)carbonyl]-2-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₈N₂O₆
MDL Number: MFCD17167330
CAS Number: 1246738-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR