1-Boc-piperidin-4-ylideneacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-Piperidin-4-ylideneacetic acid

058260-500MG 058260-1G
101914-436EA 278.46 USD
101914-436 101914-438
1-Boc-piperidin-4-ylideneacetic acid ≥95%
1-Boc-piperidin-4-ylideneacetic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MW: 241.2836 g/mol
MDL Number: MFCD04115019
CAS Number: 193085-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR