1-{2-[(tert-Butoxy)carbonyl]phenyl}-5-methyl-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

058259-500MG 058259-1G
101914-432EA 864.62 USD
101914-432 101914-434
1-{2-[(tert-Butoxy)carbonyl]phenyl}-5-methyl-1H-pyrazole-3-carboxylic acid ≥95%
1-{2-[(tert-Butoxy)carbonyl]phenyl}-5-methyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₄
MDL Number: MFCD17167309
CAS Number: 1246738-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR