1-[(tert-Butoxy)carbonyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-4-caboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminopiperidine-4-carboxylic acid, N1-BOC 2-FMOC protected

058258-500MG 058258-1G
101914-428EA 1090.77 USD
101914-428 101914-430
1-[(tert-Butoxy)carbonyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-4-caboxylic acid ≥95%
1-[(tert-Butoxy)carbonyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-4-caboxylic acid

Hazard: Irritant

Formula: C₂₆H₃₀N₂O₆
MW: 466.54 g/mol
MDL Number: MFCD17167329
CAS Number: 1246738-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR