Methyl-3-(trifluoromethyl)phenylacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(3-(trifluoromethyl)phenyl)acetate

058255-5G 058255-25G
101914-418EA 67.69 USD
101914-418 101914-420
Methyl-3-(trifluoromethyl)phenylacetate ≥95%
Methyl-3-(trifluoromethyl)phenylacetate

Hazard: Irritant

Formula: C₁₀H₉F₃O₂
MW: 218.175 g/mol
Boiling Pt: 211 °C
Density: 1.24
MDL Number: MFCD04039762
CAS Number: 62451-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR