3-(Methylcarbamoyl)propanoic acid ≥95%

Supplier: Matrix Scientific

058249-500MG 058249-1G
101914-396EA 360 USD
101914-396 101914-398
3-(Methylcarbamoyl)propanoic acid ≥95%
3-(Methylcarbamoyl)propanoic acid

Hazard: Irritant

Formula: C₅H₉NO₃
MDL Number: MFCD00020531
CAS Number: 56269-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR