6-Methoxy-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Matrix Scientific
058246-500MG
058246-1G
101914-384EA
300
USD
101914-384
101914-386
6-Methoxy-1H-pyrrolo[2,3-b]pyridine ≥95%
6-Methoxy-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₈H₈N₂O MW: 148.17 g/mol |
MDL Number:
MFCD06659665 CAS Number: 896722-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...