2-(2-Methoxyphenyl)acetohydrazide ≥95%

Supplier: Matrix Scientific

058243-500MG 058243-1G
101914-370EA 253.85 USD
101914-370 101914-372
2-(2-Methoxyphenyl)acetohydrazide ≥95%
2-(2-Methoxyphenyl)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD01610169
CAS Number: 34547-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR