2-[3-(Methoxycarbonyl)-5-methyl-1H-pyrazol-1-yl]benzoic acid ≥95%

Supplier: Matrix Scientific

058242-500MG 058242-1G
101914-366EA 573.85 USD
101914-366 101914-368
2-[3-(Methoxycarbonyl)-5-methyl-1H-pyrazol-1-yl]benzoic acid ≥95%
2-[3-(Methoxycarbonyl)-5-methyl-1H-pyrazol-1-yl]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₄
MDL Number: MFCD17167308
CAS Number: 1246738-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR