2-(2-Fluorophenyl)acetohydrazide ≥95%

Supplier: Matrix Scientific

058239-500MG 058239-1G 058239-5G
101914-350EA 84.62 USD
101914-350 101914-352 101914-354
2-(2-Fluorophenyl)acetohydrazide ≥95%
2-(2-Fluorophenyl)acetohydrazide

Hazard: Irritant

Formula: C₈H₉FN₂O
MDL Number: MFCD09729319
CAS Number: 116622-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR