4-Fluoro-5-methyl-2-nitroaniline ≥95%

Supplier: Matrix Scientific

058238-500MG 058238-1G
101914-346EA 149.23 USD
101914-346 101914-348
4-Fluoro-5-methyl-2-nitroaniline ≥95%
4-Fluoro-5-methyl-2-nitroaniline

Hazard: Irritant

Formula: C₇H₇FN₂O₂
MDL Number: MFCD17167314
CAS Number: 97389-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR