4-Fluoro-5-methylbenzene-1,2-diamine ≥95%

Supplier: Matrix Scientific

058237-500MG 058237-1G
101914-342EA 538.46 USD
101914-342 101914-344
4-Fluoro-5-methylbenzene-1,2-diamine ≥95%
4-Fluoro-5-methylbenzene-1,2-diamine

Hazard: Irritant

Formula: C₇H₉FN₂
MDL Number: MFCD17167315
CAS Number: 97389-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR