2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Fmoc-p-fluoro-DL-Phe-OH

058233-500MG 058233-1G
101914-326EA 278.46 USD
101914-326 101914-328
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid ≥95%
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid

Hazard: Irritant

Formula: C₂₄H₂₀FNO₄
MW: 405.425 g/mol
CAS Number: 264276-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR