2,4-Di-tert-butyl-5-nitrophenol ≥95%

Supplier: Matrix Scientific

058227-500MG 058227-1G
101914-304EA 292.31 USD
101914-304 101914-306
2,4-Di-tert-butyl-5-nitrophenol ≥95%
2,4-Di-tert-butyl-5-nitrophenol

Hazard: Irritant

Formula: C₁₄H₂₁NO₃
MDL Number: MFCD11052606
CAS Number: 873055-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR