3-[(Dimethylcarbamoyl)methoxy]benzoic acid ≥95%

Supplier: Matrix Scientific

058226-500MG 058226-1G
101914-300EA 375.38 USD
101914-300 101914-302
3-[(Dimethylcarbamoyl)methoxy]benzoic acid ≥95%
3-[(Dimethylcarbamoyl)methoxy]benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD08443111
CAS Number: 926259-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR