4-(Dimethylamino)but-2-enoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (2E)-4-(Dimethylamino)but-2-enoic acid

058225-500MG 058225-1G
101914-296EA 238.2 USD
101914-296 101914-298
4-(Dimethylamino)but-2-enoic acid ≥95%
4-(Dimethylamino)but-2-enoic acid

Hazard: Irritant

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD08056379
CAS Number: 98548-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR