5,7-Dichloro-3-ethylpyrazolo[1,5-a]pyrimidine ≥95%

Supplier: Matrix Scientific

058223-500MG 058223-1G
101914-286EA 767.69 USD
101914-286 101914-288
5,7-Dichloro-3-ethylpyrazolo[1,5-a]pyrimidine ≥95%
5,7-Dichloro-3-ethylpyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₈H₇Cl₂N₃
MDL Number: MFCD08448153
CAS Number: 779353-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR