5,6-Dibromo-2-chloro-1H-1,3-benzodiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Dibromo-2-chloro-1H-benzo[d]imidazole

058222-500MG 058222-1G
101914-282EA 270.77 USD
101914-282 101914-284
5,6-Dibromo-2-chloro-1H-1,3-benzodiazole ≥95%
5,6-Dibromo-2-chloro-1H-1,3-benzodiazole

Hazard: Irritant

Formula: C₇H₃Br₂ClN₂
MW: 310.39 g/mol
MDL Number: MFCD17167327
CAS Number: 142356-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR