1-(4-Cyanophenyl)guanidine nitrate ≥95%
Supplier: Matrix Scientific
058221-500MG
058221-1G
101914-278EA
270.77
USD
101914-278
101914-280
1-(4-Cyanophenyl)guanidine nitrate ≥95%
1-(4-Cyanophenyl)guanidine nitrate
Hazard: Irritant
Formula:
C₈H₉N₅O₃ |
MDL Number:
MFCD17167311 CAS Number: 5637-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...