[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methanamine hydrochloride ≥95%
Supplier: Matrix Scientific
058218-1G
058218-5G
101914-266EA
240
USD
101914-266
101914-268
[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methanamine hydrochloride ≥95%
[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methanamine hydrochloride
Hazard: Irritant
Formula:
C₇H₇Cl₂F₃N₂ |
MDL Number:
MFCD00210134 CAS Number: 175277-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...