6-Chloro-1,2,3,4-tetrahydroquinoline ≥95%

Supplier: Matrix Scientific

058217-500MG 058217-1G
101914-260EA 140 USD
101914-260 101914-262
6-Chloro-1,2,3,4-tetrahydroquinoline ≥95%
6-Chloro-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₉H₁₀ClN
MW: 167.6400 g/mol
MDL Number: MFCD09034966
CAS Number: 49716-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR