6-Chloropyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

058216-1G 058216-5G
101914-256EA 29.23 USD
101914-256 101914-258
6-Chloropyrazine-2-carboxylic acid ≥95%
6-Chloropyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
MDL Number: MFCD00837871
CAS Number: 23688-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR