N-(3-Chloro-4-methoxyphenyl)acetamide ≥95%
Supplier: Matrix Scientific
058213-500MG
058213-1G
058213-5G
101914-242EA
181.54
USD
101914-242
101914-244
101914-246
N-(3-Chloro-4-methoxyphenyl)acetamide ≥95%
N-(3-Chloro-4-methoxyphenyl)acetamide
Hazard: Irritant
Formula:
C₉H₁₀ClNO₂ |
MDL Number:
MFCD00157588 CAS Number: 7073-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...