1-(5-Chloro-4-methoxy-2-nitrophenyl)-1H-pyrrole ≥95%

Supplier: Matrix Scientific

058212-500MG 058212-1G
101914-238EA 747.69 USD
101914-238 101914-240
1-(5-Chloro-4-methoxy-2-nitrophenyl)-1H-pyrrole ≥95%
1-(5-Chloro-4-methoxy-2-nitrophenyl)-1H-pyrrole

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
MDL Number: MFCD17167328
CAS Number: 1246738-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR