N-(5-Chloro-4-methoxy-2-nitrophenyl)acetamide ≥95%

Supplier: Matrix Scientific

058211-500MG 058211-1G
101914-234EA 278.46 USD
101914-234 101914-236
N-(5-Chloro-4-methoxy-2-nitrophenyl)acetamide ≥95%
N-(5-Chloro-4-methoxy-2-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉ClN₂O₄
MDL Number: MFCD00126497
CAS Number: 160088-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR