5-Chloro-4-methoxy-2-nitroaniline ≥95%
Supplier: Matrix Scientific
058210-500MG
058210-1G
101914-230EA
389.23
USD
101914-230
101914-232
5-Chloro-4-methoxy-2-nitroaniline ≥95%
5-Chloro-4-methoxy-2-nitroaniline
Hazard: Irritant
Formula:
C₇H₇ClN₂O₃ Melting Pt: 138…139 °C |
MDL Number:
MFCD17167326 CAS Number: 160088-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...