6-Chloro-N-methoxy-N-methylpyrazine-2-carboxamide ≥95%

Supplier: Matrix Scientific

058209-500MG 058209-1G
101914-226EA 369.23 USD
101914-226 101914-228
6-Chloro-N-methoxy-N-methylpyrazine-2-carboxamide ≥95%
6-Chloro-N-methoxy-N-methylpyrazine-2-carboxamide

Hazard: Irritant

Formula: C₇H₈ClN₃O₂
MDL Number: MFCD17167334
CAS Number: 1223444-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR