N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

Synonyms: N-(6-Bromopyridin-2-yl)pivalamide

058207-500MG 058207-1G
101914-218EA 209.23 USD
101914-218 101914-220
N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide ≥95%
N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MW: 257.14 g/mol
MDL Number: MFCD08235018
CAS Number: 221087-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR