5-(Bromomethyl)-3-phenyl-1,3-oxazolidin-2-one ≥95%

Supplier: Matrix Scientific

058206-500MG 058206-1G
101914-214EA 278.46 USD
101914-214 101914-216
5-(Bromomethyl)-3-phenyl-1,3-oxazolidin-2-one ≥95%
5-(Bromomethyl)-3-phenyl-1,3-oxazolidin-2-one

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₂
MDL Number: MFCD17167320
CAS Number: 98963-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR