5-(Bromomethyl)-3-(4-fluorophenyl)-1,3-oxazolidin-2-one ≥95%

Supplier: Matrix Scientific

058204-500MG 058204-1G
101914-206EA 278.46 USD
101914-206 101914-208
5-(Bromomethyl)-3-(4-fluorophenyl)-1,3-oxazolidin-2-one ≥95%
5-(Bromomethyl)-3-(4-fluorophenyl)-1,3-oxazolidin-2-one

Hazard: Irritant

Formula: C₁₀H₉BrFNO₂
MDL Number: MFCD17167324
CAS Number: 1246738-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR