2-(2H-1,3-Benzodioxol-5-yl)acetohydrazide ≥95%

Supplier: Matrix Scientific

058201-500MG 058201-1G
101914-194EA 270.77 USD
101914-194 101914-196
2-(2H-1,3-Benzodioxol-5-yl)acetohydrazide ≥95%
2-(2H-1,3-Benzodioxol-5-yl)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MDL Number: MFCD08445398
CAS Number: 14731-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR