2-(4-Aminophenyl)butanedioic acid ≥95%

Supplier: Matrix Scientific

058200-500MG 058200-1G
101914-190EA 556.92 USD
101914-190 101914-192
2-(4-Aminophenyl)butanedioic acid ≥95%
2-(4-Aminophenyl)butanedioic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00051533
CAS Number: 22511-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR