Tetrahydro-2H-pyran-4-carbonyl chloride ≥95%
Supplier: Matrix Scientific
058109-1G
058109-5G
101914-010EA
64.62
USD
101914-010
101914-012
Tetrahydro-2H-pyran-4-carbonyl chloride ≥95%
Tetrahydro-2H-pyran-4-carbonyl chloride
Formula:
C₆H₉ClO₂ |
MDL Number:
MFCD06200863 CAS Number: 40191-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...