5H-Pyrrolo[3,2-d]pyrimidin-4-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Aminopyrrolo[3,2-d]pyrimidine
057907-5G
057907-250MG
057907-1G
101913-712EA
1227.69
USD
101913-712
102551-504
102551-506
5H-Pyrrolo[3,2-d]pyrimidin-4-amine ≥95%
5H-Pyrrolo[3,2-d]pyrimidin-4-amine
Hazard: Irritant
Formula:
C₆H₆N₄ MW: 134.14 g/mol |
MDL Number:
MFCD11520862 CAS Number: 2227-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...