5-Fluoropyrimidine ≥95%

Supplier: Matrix Scientific

057900-1G 057900-250MG
101913-694EA 692.31 USD
101913-694 102551-502
5-Fluoropyrimidine ≥95%
5-Fluoropyrimidine

Hazard: Irritant

Formula: C₄H₃FN₂
MDL Number: MFCD06658278
CAS Number: 675-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR