2-Amino-3-bromophenol ≥95%

Supplier: Matrix Scientific

057719-250MG 057719-5G
102833-234EA 101.78 USD
102833-234 102833-236
2-Amino-3-bromophenol ≥95%
2-Amino-3-bromophenol

Hazard: Irritant

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
MDL Number: MFCD08445676
CAS Number: 116435-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR