Ethyl-2-oxocyclohexanecarboxylate ≥95%

Supplier: Matrix Scientific

057587-1G 057587-5G
101913-350EA 115.38 USD
101913-350 102551-480
Ethyl-2-oxocyclohexanecarboxylate ≥95%
Ethyl-2-oxocyclohexanecarboxylate

Hazard: Irritant

Formula: C₉H₁₄O₃
MDL Number: MFCD00001631
CAS Number: 1655-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR