Ethyl-2-chlorothiazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

057465-25G 057465-5G
101913-174EA 427.69 USD
101913-174 102833-226
Ethyl-2-chlorothiazole-4-carboxylate ≥97%
Ethyl-2-chlorothiazole-4-carboxylate

Hazard: Irritant/Refrigerate

Formula: C₆H₆ClNO₂S
MDL Number: MFCD11045348
CAS Number: 41731-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR