3-Amino-2,4-dichloropyridine ≥95%

Supplier: Matrix Scientific

057417-1G 057417-25G
101913-122EA 36.92 USD
101913-122 101913-126
3-Amino-2,4-dichloropyridine ≥95%
3-Amino-2,4-dichloropyridine

Hazard: Irritant

Formula: C5H4Cl2N2
MW: 163.01 g/mol
MDL Number: MFCD09750301
CAS Number: 173772-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR