Methyl-5-chloro-2-methylnicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-chloro-2-methylnicotinate

057373-1G 057373-250MG
101913-066EA 275.38 USD
101913-066 102833-218
Methyl-5-chloro-2-methylnicotinate ≥95%
Methyl-5-chloro-2-methylnicotinate

Hazard: Irritant

Formula: C₈H₈ClNO₂
MW: 185.6100 g/mol
MDL Number: MFCD09908061
CAS Number: 350597-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR